Merck
  • Inhibition of West Nile Virus Multiplication in Cell Culture by Anti-Parkinsonian Drugs.
Inhibition of West Nile Virus Multiplication in Cell Culture by Anti-Parkinsonian Drugs.
Frontiers in microbiology (2016-03-26)
Ana B Blázquez, Miguel A Martín-Acebes, Juan-Carlos Saiz
ABSTRACT
West Nile virus (WNV) is a mosquito-borne flavivirus maintained in a transmission cycle between mosquitoes and birds, but it can also infect other vertebrates, including humans, in which it can cause neuroinvasive diseases. To date, no licensed vaccine or therapy for human use against this pathogen is yet available. A recent approach to search for new antiviral agent candidates is the assessment of long-used drugs commonly administered by clinicians to treat human disorders in drug antiviral development. In this regard, as patients with West Nile encephalitis frequently develop symptoms and features of parkinsonism, and cellular factors altered in parkinsonism, such as alpha-synuclein, have been shown to play a role on WNV infection, we have assessed the effect of four drugs (L-dopa, Selegiline, Isatin, and Amantadine), that are used as therapy for Parkinson's disease in the inhibition of WNV multiplication. L-dopa, Isatin, and Amantadine treatments significantly reduced the production of infectious virus in all cell types tested, but only Amantadine reduced viral RNA levels. These results point to antiparkinsonian drugs as possible therapeutic candidates for the development of antiviral strategies against WNV infection.
Sigma-Aldrich
Amantadine hydrochloride
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture