Merck
Reduced expression of cenp-e in human hepatocellular carcinoma.
Journal of experimental & clinical cancer research : CR (2009-12-22)
Zijie Liu, Kang Ling, Xia Wu, Ju Cao, Bin Liu, Suyan Li, Qiong Si, Yan Cai, Chen Yan, Yan Zhang, Yaguang Weng
ABSTRACT
CENP-E, one of spindle checkpoint proteins, plays a crucial role in the function of spindle checkpoint. Once CENP-E expression was interrupted, the chromosomes can not separate procedurally, and may result in aneuploidy which is a hallmark of most solid cancers, such as hepatocellular carcinoma (HCC). We investigate the expression of CENP-E in human hepatocellular carcinoma,. and analyze the effect of low CENP-E expression on chromosome separation in normal liver cell line (LO2). We determined its levels in HCC and para-cancerous tissues, human hepatocellular carcinoma-derived cell line (HepG2) and LO2 cell line using real time quantitative PCR (QPCR) and Western blot. Further to know whether reduction in CENP-E expression impairs chromosomes separation in LO2 cells. we knocked down CENP-E using shRNA expressing vector and then count the aneuploid in LO2 cells using chromosomal counts assay. We found that both CENP-E mRNA and protein levels were significantly reduced in HCC tissues and HepG2 cells compared with para-cancerous tissues and LO2 cells, respectively. A significantly-increased proportion of aneuploid in these down-knocked LO2 cells compared with those treated with control shRNA vector. Together with other results, these results reveal that CENP-E expression was reduced in human HCC tissue, and low CENP-E expression result in aneuploidy in LO2 cells.