Merck
Fluorescent cellulose aerogels containing covalently immobilized (ZnS)
Cellulose (London, England) (2013-01-01)
Huiqing Wang, Ziqiang Shao, Markus Bacher, Falk Liebner, Thomas Rosenau
ABSTRACT
Photoluminiscent (PL) cellulose aerogels of variable shape containing homogeneously dispersed and surface-immobilized alloyed (ZnS)
Sigma-Aldrich
Zinc acetate, 99.99% trace metals basis