Merck
  • Nitrile ylide dimerization: investigation of the carbene reactivity of nitrile ylides.
Nitrile ylide dimerization: investigation of the carbene reactivity of nitrile ylides.
The Journal of organic chemistry (2004-07-03)
Suzanne Fergus, Stephen J Eustace, Anthony F Hegarty
ABSTRACT
A series of novel hexaaryl diazatrienes 5 ("nitrile ylide dimers") were synthesized directly from the corresponding diaryl ketimines 12 and dichlorotoluenes 13 in a facile one-pot synthesis. The carbene character of the nitrile ylides was investigated by varying the substituents on the aromatic ring adjacent to the carbene center. The isolation of the corresponding carbene dimers as stable crystalline materials with absorption maxima (lambda(max)) from 363 to 422 nm was shown to be promoted by the absence of strongly electron-withdrawing substituents. The crystal structures indicate that the E-isomers were isolated when phenyl, 3-methylphenyl, and 3-chlorophenyl substituents are present at the carbene carbon; the Z-isomer was isolated when the more sterically hindered 2,4,6-trimethylphenyl substituent (Mes) is present. The (1)H NMR spectra of the E-isomers demonstrate the nonequivalence of the aromatic rings, in which two of the aromatic rings of the imine moiety are pseudoaxial and the remaining aromatic rings are pseudoequatorial. The reactions proceed via the intermediate nitrile ylides 1 generated by the base-promoted 1,1-elimination of HCl from the intermediate chloroimine 14. The nitrile ylide was also generated by 1,3-elimination of HCl from the imidoyl chloride 18, confirming common pathways via the nitrile ylide as the dimer products obtained from these different routes were identical. The strongly electron-withdrawing 4-nitrophenyl substituent promotes the linear carbanion character of the 1,3-dipole and no dimer is formed.
Sigma-Aldrich
Benzophenone imine, 95%
Pricing and availability is not currently available.