Merck
  • Relationship between interleukin-1 type 1 and 2 receptor gene polymorphisms and the expression level of membrane-bound receptors.
Relationship between interleukin-1 type 1 and 2 receptor gene polymorphisms and the expression level of membrane-bound receptors.
Cellular & molecular immunology (2014-07-01)
F F Vasilyev, A N Silkov, S V Sennikov
ABSTRACT
The biological activity of the multifunctional cytokine interleukin-1 (IL-1) is mediated by its receptors. The aim of this study was to determine if an association exists between single nucleotide polymorphisms (SNPs) in the IL-1 type 1 and 2 receptor genes (IL1R1 and IL1R2) and the expression level of membrane-bound IL1Rs on subpopulations of mononuclear cells or serum levels of soluble IL-1 receptors. It was observed that healthy individuals with the genotype TT in SNP rs2234650:C>T had a lower percentage of intact CD14(+) monocytes expressing IL1R1 on their surface. The SNP rs4141134:T>C in IL1R2 has also been associated with the percentage of intact CD3(+) T cells expressing IL1R2. Furthermore, individuals carrying the CC allele of SNP rs4141134:T>C and the TT allele of SNP rs2071008:T>G in IL1R2 had a lower density of IL1R2s on the surface of CD14(+) monocytes in lipopolysaccharide (LPS)-stimulated PBMC cultures. In summary, this study demonstrated that IL-1 receptor gene polymorphisms could be one of the factors influencing the expression of membrane-bound IL-1 receptors (IL1R) on immunocompetent cells.
Sigma-Aldrich
Interleukin-1 Soluble Receptor, Type I human, >97% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder