Merck
  • Chemical derivatization of neurosteroids for their trace determination in sea lamprey by UPLC-MS/MS.
Chemical derivatization of neurosteroids for their trace determination in sea lamprey by UPLC-MS/MS.
Talanta (2016-01-01)
Ugo Bussy, Mar Huertas, Yu-Wen Chung-Davidson, Ke Li, Weiming Li
ABSTRACT
This article describes the development and validation of a sensitive and robust method for the determination of neurosteroids in sea lamprey, an ancestral animal in vertebrate evolution. Chemical derivatization was used to enhance the detection of neurosteroids containing ketone function by ultra-high performance liquid chromatography tandem mass spectrometry (UPLC-MS/MS). Iminooxy derivatives of 12 oxosteroids and three internal standards were monitored by positive electrospray tandem mass spectrometry using the neutral loss of sulfate. Limit of quantification, extraction recovery and matrix effect were first evaluated and SPE using C18 sorbent was selected after comparison with liquid/liquid extraction and protein precipitation. Matrix effect ranged from 89.6% to 113.1% in plasma and from 79.8% to 100.0% in the brain. Recovery values ranged from 80.0% to 103.8% in plasma and from 86.3% to 107.9% in the brain. Chromatographic separation was achieved by reverse phase chromatography (2.1mm×100mm, 1.7µm particle size, C18) with a binary gradient between methanol and 0.1% formic acid in water. Limit of quantification ranged from 5 to 10pg/mL and was up to 80 times lower than those for non-derivatized steroids. Accuracy and precision parameters were determined and inter- and intra-day at three concentrations: 50, 500 and 5000pg/mL. This method was applied to analyze real samples. progesterone (P), pregnenolone (P5), 7-hydroxy-pregnenolpne (7P5), 17-hydroxy-pregnenolpne (17P5)dehydroepiandrosterone (DHEA), androstenedienone (A4), testosterone (T), dihydrotestosterone (DHT), allopregnanolone (THP), 11-hydroxy-androstenedienone (11A4) and 11-Deoxycortisol (S) were measured in sea lamprey brain and plasma matrixes.