Merck
  • Microwave-assisted preparation of dialkylimidazolium tetrachloroaluminates and their use as catalysts in the solvent-free tetrahydropyranylation of alcohols and phenols.
Microwave-assisted preparation of dialkylimidazolium tetrachloroaluminates and their use as catalysts in the solvent-free tetrahydropyranylation of alcohols and phenols.
Chemical communications (Cambridge, England) (2002-07-18)
Vasudevan V Namboodiri, Rajender S Varma
ABSTRACT
Microwave-assisted preparation of dialkylimidazolium tetrachloroaluminates, [CnMIM][AlCl4] and their application as recyclable catalysts for the efficient and eco-friendly protection of alcohols as tetrahydropyranyl (THP) ethers are described. The same catalyst can also be utilized for the deprotection of THP ethers.
Sigma-Aldrich
1-Hexyl-3-methylimidazolium chloride, ≥97.0% (HPLC)