Merck
  • Hot-melt extrusion of polyvinyl alcohol for oral immediate release applications.
Hot-melt extrusion of polyvinyl alcohol for oral immediate release applications.
International journal of pharmaceutics (2015-07-15)
W De Jaeghere, T De Beer, J Van Bocxlaer, J P Remon, C Vervaet
ABSTRACT
The primary purpose of this study was to process partially hydrolyzed PVOH grades (degree of hydroxylation (DH): 33-88%) via HME and to evaluate them as carrier for oral immediate release dosage forms in order to improve the release rate of poorly water soluble drugs (i.e., HCT and CEL) via the formulation of solid dispersions. PVOH grades (DH >70%) were able to solubilize HCT and CEL up to 15%, but required higher extrusion temperature, due to the crystalline nature of PVOH. The highest drug release rate was observed from hot-melt extruded PVOH samples with a high DH. While drug release from extrudates consisting of PVOH with a low DH was affected by ionic strength, there was no influence of pH and ionic strength on HCT release from PVOH samples with a higher DH. However, PVOH (DH >70%) required higher extrusion temperatures, which could hamper its application for thermosensitive drugs. Therefore, the secondary purpose was to investigate the effect of sorbitol, a water-soluble plasticizer, on the thermal properties of hot-melt extruded PVOH (DH >70%). The melting of PVOH/sorbitol mixture was required to establish molecular interactions between PVOH and sorbitol. These molecular interactions were reflected in the HME behavior: whereas an extrusion temperature of 180 °C was necessary to process physical mixtures of PVOH (DH >70%) and sorbitol, only 140 °C was necessary during re-extrusion (after quench cooling and cryomilling) of the PVOH/sorbitol mixture. In addition, the in vitro and in vivo dug release of plasticized PVOH was examined; whereas the CEL/PVO/sorbitol system was able to maintain supersaturation during in vitro dissolution (0.1N HCl) compared to Celebrex(®), the in vivo pharmacokinetic parameters (AUC0-24h, Cmax and Tmax) were highly comparable.
Sigma-Aldrich
Potassium fluoride on Celite®, extent of labeling: 50 wt. % loading, technical grade
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), Molecular Biology
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
Potassium fluoride, BioUltra, ≥99.5% (F)
Sigma-Aldrich
Potassium fluoride, anhydrous, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Potassium fluoride, ≥99.97% trace metals basis