Merck
  • D-bifunctional protein deficiency with fetal ascites, polyhydramnios, and contractures of hands and toes.
D-bifunctional protein deficiency with fetal ascites, polyhydramnios, and contractures of hands and toes.
The Journal of pediatrics (2001-12-18)
K Nakano, Z Zhang, N Shimozawa, N Kondo, N Ishii, M Funatsuka, S Shirakawa, M Itoh, S Takashima, M Une, R R Kana-aki, K Mukai, M Osawa, Y Suzuki
ABSTRACT
Fetal abnormalities including chylous ascites, polyhydramnios, claw hands, and hammer toes were identified in an infant who had a missense mutation R106P and a 52bp deletion in the gene for a peroxisomal beta-oxidation enzyme, D-3-hydroxyacyl-CoA dehydratase/D-3-hydroxyacyl-CoA dehydrogenase, D-bifunctional protein. The patient had psychomotor retardation and craniofacial dysmorphism and died at 7 months of age. The patient had atypical fetal manifestations of this enzyme deficiency.