Merck
  • Expression of the hypoxia-inducible monocarboxylate transporter MCT4 is increased in triple negative breast cancer and correlates independently with clinical outcome.
Expression of the hypoxia-inducible monocarboxylate transporter MCT4 is increased in triple negative breast cancer and correlates independently with clinical outcome.
Biochemical and biophysical research communications (2014-07-25)
J Doyen, C Trastour, F Ettore, I Peyrottes, N Toussant, J Gal, K Ilc, D Roux, S K Parks, J M Ferrero, J Pouysségur
ABSTRACT
(18)Fluor-deoxy-glucose PET-scanning of glycolytic metabolism is being used for staging in many tumors however its impact on prognosis has never been studied in breast cancer. Glycolytic and hypoxic markers: glucose transporter (GLUT1), carbonic anhydrase IX (CAIX), monocarboxylate transporter 1 and 4 (MCT1, 4), MCT accessory protein basigin and lactate-dehydrogenase A (LDH-A) were assessed by immunohistochemistry in two cohorts of breast cancer comprising 643 node-negative and 127 triple negative breast cancers (TNBC) respectively. In the 643 node-negative breast tumor cohort with a median follow-up of 124 months, TNBC were the most glycolytic (≈70%), followed by Her-2 (≈50%) and RH-positive cancers (≈30%). Tumoral MCT4 staining (without stromal staining) was a strong independent prognostic factor for metastasis-free survival (HR=0.47, P=0.02) and overall-survival (HR=0.38, P=0.002). These results were confirmed in the independent cohort of 127 cancer patients. Glycolytic markers are expressed in all breast tumors with highest expression occurring in TNBC. MCT4, the hypoxia-inducible lactate/H(+) symporter demonstrated the strongest deleterious impact on survival. We propose that MCT4 serves as a new prognostic factor in node-negative breast cancer and can perhaps act soon as a theranostic factor considering the current pharmacological development of MCT4 inhibitors.