Merck
  • Mycobacterium tuberculosis lineage 7 strains are associated with prolonged patient delay in seeking treatment for pulmonary tuberculosis in Amhara Region, Ethiopia.
Mycobacterium tuberculosis lineage 7 strains are associated with prolonged patient delay in seeking treatment for pulmonary tuberculosis in Amhara Region, Ethiopia.
Journal of clinical microbiology (2015-02-13)
Solomon A Yimer, Gunnstein Norheim, Amine Namouchi, Ephrem D Zegeye, Wibeke Kinander, Tone Tønjum, Shiferaw Bekele, Turid Mannsåker, Gunnar Bjune, Abraham Aseffa, Carol Holm-Hansen
ABSTRACT
Recent genotyping studies of Mycobacterium tuberculosis in Ethiopia have reported the identification of a new phylogenetically distinct M. tuberculosis lineage, lineage 7. We therefore investigated the genetic diversity and association of specific M. tuberculosis lineages with sociodemographic and clinical parameters among pulmonary TB patients in the Amhara Region, Ethiopia. DNA was isolated from M. tuberculosis-positive sputum specimens (n=240) and analyzed by PCR and 24-locus mycobacterial interspersed repetitive unit-variable-number tandem-repeat (MIRU-VNTR) analysis and spoligotyping. Bioinformatic analysis assigned the M. tuberculosis genotypes to global lineages, and associations between patient characteristics and genotype were evaluated using logistic regression analysis. The study revealed a high diversity of modern and premodern M. tuberculosis lineages, among which approximately 25% were not previously reported. Among the M. tuberculosis strains (n=138) assigned to seven subgroups, the largest cluster belonged to the lineage Central Asian (CAS) (n=60; 26.0%), the second largest to lineage 7 (n=36; 15.6%), and the third largest to the lineage Haarlem (n=35; 15.2%). Four sublineages were new in the MIRU-VNTRplus database, designated NW-ETH3, NW-ETH1, NW-ETH2, and NW-ETH4, which included 24 (10.4%), 18 (7.8%), 8 (3.5%), and 5 (2.2%) isolates, respectively. Notably, patient delay in seeking treatment was significantly longer among patients infected with lineage 7 strains (Mann-Whitney test, P<0.008) than in patients infected with CAS strains (adjusted odds ratio [AOR], 4.7; 95% confidence interval [CI], 1.6 to 13.5). Lineage 7 strains also grew more slowly than other M. tuberculosis strains. Cases of Haarlem (OR, 2.8; 95% CI, 1.2 to 6.6) and NW-ETH3 (OR, 2.8; 95% CI, 1.0 to 7.3) infection appeared in defined clusters. Intensified active case finding and contact tracing activities in the study region are needed to expedite diagnosis and treatment of TB.
Product Description
Amphotericin B, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nalidixic acid, ≥98%
Nalidixic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Nalidixic acid, analytical standard
Trimethoprim for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Supelco
Trimethoprim, VETRANAL®, analytical standard
Sigma-Aldrich
Trimethoprim, ≥98.5%
Supelco
Trimethoprim, Pharmaceutical Secondary Standard; Certified Reference Material
Trimethoprim, European Pharmacopoeia (EP) Reference Standard