Merck
  • Merlin deficiency predicts FAK inhibitor sensitivity: a synthetic lethal relationship.
Merlin deficiency predicts FAK inhibitor sensitivity: a synthetic lethal relationship.
Science translational medicine (2014-05-23)
Irina M Shapiro, Vihren N Kolev, Christian M Vidal, Yuwaraj Kadariya, Jennifer E Ring, Quentin Wright, David T Weaver, Craig Menges, Mahesh Padval, Andrea I McClatchey, Qunli Xu, Joseph R Testa, Jonathan A Pachter
ABSTRACT
The goal of targeted therapy is to match a selective drug with a genetic lesion that predicts for drug sensitivity. In a diverse panel of cancer cell lines, we found that the cells most sensitive to focal adhesion kinase (FAK) inhibition lack expression of the neurofibromatosis type 2 (NF2) tumor suppressor gene product, Merlin. Merlin expression is often lost in malignant pleural mesothelioma (MPM), an asbestos-induced aggressive cancer with limited treatment options. Our data demonstrate that low Merlin expression predicts for increased sensitivity of MPM cells to a FAK inhibitor, VS-4718, in vitro and in tumor xenograft models. Disruption of MPM cell-cell or cell-extracellular matrix (ECM) contacts with blocking antibodies suggests that weak cell-cell adhesions in Merlin-negative MPM cells underlie their greater dependence on cell-ECM-induced FAK signaling. This provides one explanation of why Merlin-negative cells are vulnerable to FAK inhibitor treatment. Furthermore, we validated aldehyde dehydrogenase as a marker of cancer stem cells (CSCs) in MPM, a cell population thought to mediate tumor relapse after chemotherapy. Whereas pemetrexed and cisplatin, standard-of-care agents for MPM, enrich for CSCs, FAK inhibitor treatment preferentially eliminates these cells. These preclinical results provide the rationale for a clinical trial in MPM patients using a FAK inhibitor as a single agent after first-line chemotherapy. With this design, the FAK inhibitor could potentially induce a more durable clinical response through reduction of CSCs along with a strong antitumor effect. Furthermore, our data suggest that patients with Merlin-negative tumors may especially benefit from FAK inhibitor treatment.