Merck
  • Withdrawal from long-term methamphetamine self-administration 'normalizes' neurometabolites in rhesus monkeys: a (1) H MR spectroscopy study.
Withdrawal from long-term methamphetamine self-administration 'normalizes' neurometabolites in rhesus monkeys: a (1) H MR spectroscopy study.
Addiction biology (2013-08-06)
Shaolin Yang, Annabelle M Belcher, Svetlana Chefer, D Bruce Vaupel, Charles W Schindler, Elliot A Stein, Yihong Yang
ABSTRACT
(1) H magnetic resonance spectroscopy has demonstrated alterations in several neurometabolites in methamphetamine (METH)-dependent individuals in brain regions implicated in addiction. Yet, it is unclear whether these neurochemicals return to homeostatic levels after an individual abstains from drug use, a difficult question to address due to high recidivism and poor study retention in human subjects. We thus utilized a non-human primate model of addiction to explore the effects of long-term drug exposure and withdrawal on brain neurochemistry. Ten rhesus macaque monkeys on an active METH self-administration protocol (average use 4.6 ± 0.8 years, average daily intake between 0.4 and 1.2 mg/kg) and 10 age- and sex-matched drug-naive controls (CONT) served as subjects. Concentrations of several neurochemicals were evaluated at several timepoints following withdrawal from drug availability (10 monkeys at 1 week and 1 and 3 months, and 6 monkeys at 6 and 12 months; CONT examined at one timepoint). At 1 week following METH withdrawal, we found increases in myo-inositol in anterior cingulate cortex in the METH group relative to CONT. These alterations showed a linear pattern of decreased levels ('normalization') by 1 year of abstinence. We also found decreases in glutamine and Glx (composed mainly of glutamate and glutamine) in the caudate-putamen of the same animals at early withdrawal that showed a similar linear pattern of increasing concentration by 1 year. These results demonstrate that despite protracted, long-term use, neurochemical changes seen following long-term drug administration do not persist following prolonged abstinence, suggesting therapeutic effects of long-term withdrawal from drug use.
Product Description
Supelco
(+)-Methamphetamine hydrochloride solution, analytical standard, for drug analysis, 1.0 mg/mL in methanol
Sigma-Aldrich
(+)-Methamphetamine hydrochloride