- Silver-Mediated Synthesis of Indolizines via Oxidative C-H functionalization and
Silver-Mediated Synthesis of Indolizines via Oxidative C-H functionalization and
An efficient strategy for the synthesis of indolizines from readily available starting materials via oxidative C-H functionalization and