Merck
  • Expression and activation of TGF-beta isoforms in acute allergen-induced remodelling in asthma.
Expression and activation of TGF-beta isoforms in acute allergen-induced remodelling in asthma.
Thorax (2007-01-26)
Alfons Torrego, Mark Hew, Tim Oates, Maria Sukkar, Kian Fan Chung
ABSTRACT
Airway wall remodelling and inflammation are features of chronic asthma. Transforming growth factor beta (TGF-beta) has been implicated in these processes. To determine the effect of allergen challenge on airway inflammation and remodelling and whether TGF-beta isoforms and the Smad signalling pathways are involved. Thirteen patients with atopic asthma underwent inhalational challenge with 0.9% saline, followed by allergen 3-4 weeks later. After both challenges, fibreoptic bronchoscopy was undertaken to obtain bronchial biopsies and tissue samples were processed for immunohistochemistry and examined by microscopy. Forced expiratory volume in 1 s (FEV(1)) fell after allergen challenge (mean (SE) -28.1 (0.9)% at 30 min with a late response at 7 hours (-23.0 (1.2)%). Allergen challenge caused an increase in neutrophils and eosinophils in the bronchial mucosa compared with saline. Sub-basement membrane (SBM) thickness did not change after allergen, but tenascin deposition in SBM was increased. Intranuclear (activated) Smad 2/3 and Smad 4 detected by immunohistochemistry were increased after allergen challenge in epithelial and subepithelial cells of bronchial biopsies. No inhibitory Smad (Smad 7) protein was detected. TGF-beta isoforms 1, 2 and 3 were expressed predominantly in bronchial epithelium after saline and allergen challenges, but only TGF-beta(2) expression was increased after allergen. Double immunostaining showed an increase in TGF-beta(2) positive eosinophils and neutrophils but not in TGF-beta(1) positive eosinophils and neutrophils after allergen challenge. TGF-beta(2) may contribute to the remodelling changes in allergic asthma following single allergen exposure.
Sigma-Aldrich
3,3′-Diaminobenzidine tetrahydrochloride, tablet, 10 mg substrate per tablet