Merck
  • Cloning and expression of a novel cysteine-rich secreted protein family member expressed in thyroid and pancreatic mesoderm within the chicken embryo.
Cloning and expression of a novel cysteine-rich secreted protein family member expressed in thyroid and pancreatic mesoderm within the chicken embryo.
Mechanisms of development (2001-04-05)
D M Smith, L A Collins-Racie, V A Marigo, D J Roberts, N M Davis, C Hartmann, R Schweitzer, E R LaVallie, L Gamer, J McCoy, C J Tabin
ABSTRACT
We have isolated a new chicken gene that is a member of the cysteine-rich secreted protein family (CRISP). The CRISP family is composed of over 70 members that are found in many phyla of organisms, including: vertebrates, plants, fungi, yeast, and insects. Here we describe the cloning of a novel member of this family, SugarCrisp, and its expression pattern throughout chicken embryogenesis. We also describe its utility as a marker of thyroid and pancreatic mesoderm in the developing chicken embryo and its expression within the human and mouse in glandular tissue.