Merck
  • Cellobiose dehydrogenase from the ligninolytic basidiomycete Ceriporiopsis subvermispora.
Cellobiose dehydrogenase from the ligninolytic basidiomycete Ceriporiopsis subvermispora.
Applied and environmental microbiology (2009-03-10)
Wolfgang Harreither, Christoph Sygmund, Evelyn Dünhofen, Rafael Vicuña, Dietmar Haltrich, Roland Ludwig
ABSTRACT
Cellobiose dehydrogenase (CDH), an extracellular flavocytochrome produced by several wood-degrading fungi, was detected in cultures of the selective delignifier Ceriporiopsis subvermispora when grown on a cellulose- and yeast extract-based liquid medium. CDH amounted to up to 2.5% of total extracellular protein during latter phases of the cultivation and thus suggested an important function for the fungus under the given conditions. The enzyme was purified 44-fold to apparent homogeneity. It was found to be present in two glycoforms of 98 kDa and 87 kDa with carbohydrate contents of 16 and 4%, respectively. The isoelectric point of both glycoforms is around 3.0, differing by 0.1 units, which is the most acidic value so far reported for a CDH. By using degenerated primers of known CDH sequences, one cdh gene was found in the genomic DNA, cloned, and sequenced. Alignment of the 774-amino-acid protein sequence revealed a high similarity to CDH from other white rot fungi. One notable difference was found in the longer interdomain peptide linker, which might affect the interdomain electron transfer at higher temperatures. The preferred substrate of C. subvermispora CDH is cellobiose, while glucose conversion is strongly discriminated by a 155,000-fold-lower catalytic efficiency. This is a typical feature of a basidiomycete CDH, as are the acidic pH optima for all tested electron acceptors in the range from 2.5 to 4.5.