Merck
  • Epigenetic priming of memory updating during reconsolidation to attenuate remote fear memories.
Epigenetic priming of memory updating during reconsolidation to attenuate remote fear memories.
Cell (2014-01-21)
Johannes Gräff, Nadine F Joseph, Meryl E Horn, Alireza Samiei, Jia Meng, Jinsoo Seo, Damien Rei, Adam W Bero, Trongha X Phan, Florence Wagner, Edward Holson, Jinbin Xu, Jianjun Sun, Rachael L Neve, Robert H Mach, Stephen J Haggarty, Li-Huei Tsai
ABSTRACT
Traumatic events generate some of the most enduring forms of memories. Despite the elevated lifetime prevalence of anxiety disorders, effective strategies to attenuate long-term traumatic memories are scarce. The most efficacious treatments to diminish recent (i.e., day-old) traumata capitalize on memory updating mechanisms during reconsolidation that are initiated upon memory recall. Here, we show that, in mice, successful reconsolidation-updating paradigms for recent memories fail to attenuate remote (i.e., month-old) ones. We find that, whereas recent memory recall induces a limited period of hippocampal neuroplasticity mediated, in part, by S-nitrosylation of HDAC2 and histone acetylation, such plasticity is absent for remote memories. However, by using an HDAC2-targeting inhibitor (HDACi) during reconsolidation, even remote memories can be persistently attenuated. This intervention epigenetically primes the expression of neuroplasticity-related genes, which is accompanied by higher metabolic, synaptic, and structural plasticity. Thus, applying HDACis during memory reconsolidation might constitute a treatment option for remote traumata.
Sigma-Aldrich
Histone Deacetylase 3 (HDAC3) Inhibitor Screening Kit, 100 assays in 96 well plates
Sigma-Aldrich
Histone Deacetylase 3 (HDAC3) Activity Assay Kit, 100 assays in 96 well plates
Sigma-Aldrich
Histone Deacetylase 8 (HDAC8) Activity Assay Kit, 100 assays in 96 well plates