Merck
  • Simple iron-amine catalysts for the cross-coupling of aryl Grignards with alkyl halides bearing beta-hydrogens.
Simple iron-amine catalysts for the cross-coupling of aryl Grignards with alkyl halides bearing beta-hydrogens.
Chemical communications (Cambridge, England) (2005-08-16)
Robin B Bedford, Duncan W Bruce, Robert M Frost, Michael Hird
ABSTRACT
Mixtures of iron(III) chloride and appropriate amine ligands are active catalysts for the coupling of aryl Grignard reagents with primary and secondary alkyl halide substrates bearing beta-hydrogens, under mild and simple reaction conditions.
Sigma-Aldrich
(Bromomethyl)cyclopropane, 97%