Merck
Albocycline: structure determination by x-ray crystallography.
The Journal of antibiotics (1982-12-01)
R C Thomas, C G Chidester
ABSTRACT
The structure and absolute configuration of the macrolide antibiotic albocycline (1a) has been determined by X-ray crystallographic analysis on the derived p-bromobenzoate (1b). The absolute configuration of albocycline is 4R, 7S, 12S, 13R.
Sigma-Aldrich
Albocycline, ≥95% (LC/MS-ELSD)