Merck
  • Microparticles for delivering therapeutic peptides and proteins to the lumen of the small intestine.
Microparticles for delivering therapeutic peptides and proteins to the lumen of the small intestine.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2001-07-05)
M J Amorim, J P Ferreira
ABSTRACT
Several different peptides and proteins, such as the pancreatic trypsin inhibitor, growth factors and trefoil peptides, are known to play important roles in maintaining the structure and function of the gastrointestinal wall. With the advent of recombinant biotechnology, it has become feasible to test some of these proteins as therapeutics in different inflammatory conditions of the intestines. However, the harsh pH and enzymatic conditions of the stomach can lead to their inactivation. This research was aimed at the development of particulate, gastric-resistant pharmaceutical forms, incorporating those bioactive molecules. Mixtures of proteins in powder form were coated with cellulose acetate phthalate, Eudragit S100 or Eudragit RS PO, using simple preparation techniques based on single emulsion/solvent evaporation. Using aprotinin as a model drug, it was found that these procedures were effective in microencapsulating protein in the solid form without affecting its biological activity. Furthermore, and in particular with the first two polymers above, particles showed adequate in vitro release patterns for the applications envisioned.
Sigma-Aldrich
Magnesium stearate, puriss., meets analytical specification of Ph. Eur., BP, ≥90% stearic and palmitic acid basis, ≥40% stearic acid basis (GC), 4.0-5.0% Mg basis (calc on dry sub.)
Pricing and availability is not currently available.