Merck
  • Asymmetric synthesis of 8-aminoindolizidine from chiral 2-pyrroleimines.
Asymmetric synthesis of 8-aminoindolizidine from chiral 2-pyrroleimines.
The Journal of organic chemistry (2008-10-02)
Vincenzo Giulio Albano, Andrea Gualandi, Magda Monari, Diego Savoia
ABSTRACT
1-Allyl-2-pyrroleimines obtained from (S)-valinol and (S)-phenylglycinol underwent highly diastereoselective addition of allylmagnesium chloride, used in excess amounts, to give the corresponding secondary amines with concomitant allyl to (Z)-1-propenyl isomerization of the 1-pyrrole substituent. Transformation of the 2-amino alcohol moiety to an oxazolidinone, or its cleavage and subsequent N-protection, followed by ring-closing metathesis of the two alkene groups gave the unsaturated bicyclic compound. Full hydrogenation of the alkene function and the aromatic rings afforded the indolizidine derivative as a mixture of two or three diastereomers with a ratio which was dependent on the nature of both the N-substituent and the catalyst. The two prevalent diastereomers were isolated, and their configuration was determined by X-ray crystallographic analysis.
Sigma-Aldrich
2-Amino-3-methyl-1-butanol, 97%