Merck
  • Single step optical fabrication of a DFB laser device in fluorescent azobenzene-containing materials.
Single step optical fabrication of a DFB laser device in fluorescent azobenzene-containing materials.
Advanced materials (Deerfield Beach, Fla.) (2012-06-01)
Leonid M Goldenberg, Victor Lisinetskii, Yuri Gritsai, Joachim Stumpe, Sigurd Schrader
ABSTRACT
Distributed feedback (DFB) lasers are produced directly in fluorescent azobenzene-containing materials using a single holographic optical step. Surface relief grating capable of producing images in fluorescence microscopy can be holographically formed in a number of materials.
Sigma-Aldrich
Difluoro{3-ethyl-5-[1-(4-ethyl-3,5-dimethyl-2H-pyrrol-2-ylidene-N)ethyl]-2,4-dimethyl-1H-pyrrolato-N}boron, 98% (HPLC)