Merck
  • Rapid and selective catalytic oxidation of secondary alcohols at room temperature by using (N-heterocyclic carbene)-ni(0) systems.
Rapid and selective catalytic oxidation of secondary alcohols at room temperature by using (N-heterocyclic carbene)-ni(0) systems.
Chemistry (Weinheim an der Bergstrasse, Germany) (2010-05-11)
Christophe Berini, Ole H Winkelmann, Jennifer Otten, David A Vicic, Oscar Navarro
ABSTRACT
The selective, anaerobic catalytic oxidation of secondary alcohols at room temperature by using an in situ (N-heterocyclic carbene)-Ni(0) system is presented. The use of non-anhydrous, non-degassed 2,4-dichlorotoluene as both the oxidant and the solvent allows for very short reaction times and very high yields. In addition, a well-defined (N-heterocyclic carbene)-Ni(0) complex was synthesized and applied to these oxidation reactions.
Sigma-Aldrich
2,4-Dichlorotoluene, 99%
Pricing and availability is not currently available.