Merck
  • Selective methylative homologation: an alternate route to alkane upgrading.
Selective methylative homologation: an alternate route to alkane upgrading.
Journal of the American Chemical Society (2008-08-14)
John E Bercaw, Nilay Hazari, Jay A Labinger, Valerie J Scott, Glenn J Sunley
ABSTRACT
InI3 catalyzes the reaction of branched alkanes with methanol to produce heavier and more highly branched alkanes, which are more valuable fuels. The reaction of 2,3-dimethylbutane with methanol in the presence of InI3 at 180-200 degrees C affords the maximally branched C7 alkane, 2,2,3-trimethylbutane (triptane). With the addition of catalytic amounts of adamantane the selectivity of this transformation can be increased up to 60%. The lighter branched alkanes isobutane and isopentane also react with methanol to generate triptane, while 2-methylpentane is converted into 2,3-dimethylpentane and other more highly branched species. Observations implicate a chain mechanism in which InI3 activates branched alkanes to produce tertiary carbocations which are in equilibrium with olefins. The latter react with a methylating species generated from methanol and InI3 to give the next-higher carbocation, which accepts a hydride from the starting alkane to form the homologated alkane and regenerate the original carbocation. Adamantane functions as a hydride transfer agent and thus helps to minimize competing side reactions, such as isomerization and cracking, that are detrimental to selectivity.
Sigma-Aldrich
2,2,3-Trimethylbutane, ≥99%
Pricing and availability is not currently available.