Merck
  • Pericyclic reaction of a zwitterionic salt of an enedione-diazoester. A novel strategy for the synthesis of highly functionalized resorcinols.
Pericyclic reaction of a zwitterionic salt of an enedione-diazoester. A novel strategy for the synthesis of highly functionalized resorcinols.
Organic letters (2010-09-03)
Yu Liu, Kanwarpal Bakshi, Peter Zavalij, Michael P Doyle
ABSTRACT
Enedione-diazoesters formed from 3-TBSO-2-diazo-3-butenoates undergo base-catalyzed pericyclization that with dinitrogen extrusion and methyl migration provide a novel and efficient route to 2-carboalkoxyresorcinols. Intercepting the intermediate enolate anion with methyl vinyl ketone leads to the corresponding 4-substituted 2-carboalkoxyresorcinol and suggests generalization of this methodology.
Sigma-Aldrich
trans-4-Methoxy-3-buten-2-one, technical grade, 90%