Merck
  • Copolymerization of ethylene with sterically hindered 3,3-dimethyl-1-butene using a chain-walking pd-diimine catalyst.
Copolymerization of ethylene with sterically hindered 3,3-dimethyl-1-butene using a chain-walking pd-diimine catalyst.
Macromolecular rapid communications (2011-05-19)
Peng Xiang, Zhibin Ye
ABSTRACT
In this Communication, the copolymerization of ethylene with a sterically hindered α-olefin comonomer, γ-trisubstituted 3,3-dimethyl-1-butene (DMB), using a chain-walking Pd-diimine catalyst, [(ArNC(Me)(Me)CNAr)Pd(CH(3) )(NCMe)]SbF(6) (Ar2,6-(iPr)(2) C(6) H(3) ) (1) is reported. In spite of its high steric bulkiness in the close proximity of the double bond, appreciable DMB incorporations (up to 3 mol-%) are successfully achieved in the copolymers. The chain microstructure of the copolymers is elucidated, and the effect of DMB incorporation on polymer topology and thermal properties are examined. This work thus demonstrates the high capability of the Pd-diimine catalyst in incorporating sterically encumbered α-olefins.
Sigma-Aldrich
3,3-Dimethyl-1-butene, ≥97% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
3,3-Dimethyl-1-butene, ≥98.5% (GC)
Pricing and availability is not currently available.