Merck
  • Critical review of dietary caffeine and blood pressure: a relationship that should be taken more seriously.
Critical review of dietary caffeine and blood pressure: a relationship that should be taken more seriously.
Psychosomatic medicine (2004-01-30)
Jack E James
ABSTRACT
This review aims to determine whether, and to what extent, dietary caffeine may be a risk to cardiovascular health. A critical review of the relevant experimental and epidemiologic literature was conducted, with particular reference to studies of caffeine and blood pressure (BP). There is extensive evidence that caffeine at dietary doses increases BP. However, concern that the drug may contribute to cardiovascular disease appears to have been dampened by (1) the belief that habitual use leads to the development of tolerance, and (2) confusion regarding relevant epidemiologic findings. When considered comprehensively, findings from experimental and epidemiologic studies converge to show that BP remains reactive to the pressor effects of caffeine in the diet. Overall, the impact of dietary caffeine on population BP levels is likely to be modest, probably in the region of 4/2 mm Hg. At these levels, however, population studies of BP indicate that caffeine use could account for premature deaths in the region of 14% for coronary heart disease and 20% for stroke. Current evidence supports the conclusion that the BP-elevating effects of dietary caffeine may be contributing appreciably to population levels of cardiovascular mortality and morbidity. Accordingly, strategies for encouraging reduced dietary levels of caffeine deserve serious consideration.
Product Description
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Caffeine, European Pharmacopoeia (EP) Reference Standard
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard