Merck
  • [Extrapyramidal toxicity caused by metoclopramide and clebopride: study of voluntary notifications of adverse effects to the Spanish Drug Surveillance System].
[Extrapyramidal toxicity caused by metoclopramide and clebopride: study of voluntary notifications of adverse effects to the Spanish Drug Surveillance System].
Atencion primaria (1998-06-03)
R Cuena Boy, M A Maciá Martínez
ABSTRACT
To clarify if there is any basis for the hypothesis that Clebopride leads to more extrapyramidal reactions than Metoclopramide. Observational, longitudinal, retrospective and comparative study of two series of cases. The entire Spanish healthcare system. Those notified to the Spanish Drug watch system as possibly having suffered an adverse reaction to Metoclopramide (n = 98) or Clebopride (n = 123) between 1/1/1990 and 10/6/1997. None. 84.3% of suspected adverse reactions to Clebopride and 51.6% of those to Metoclopramide had a non-hospital precedence (P < 0.001). In 48.0% of suspected adverse reactions to Metoclopramide and 72.4% of those to Clebopride, there was extrapyramidal toxicity (P = 0.021). There is a basis for the hypothesis that Clebopride causes more extrapyramidal reactions than Metoclopramide. It was reasonable to realize a study based on this hypothesis.