Merck
  • Degradation of bradykinin in human urine by carboxypeptidase Y-like exopeptidase and neutral endopeptidase and their inhibition by ebelactone B and phosphoramidon.
Degradation of bradykinin in human urine by carboxypeptidase Y-like exopeptidase and neutral endopeptidase and their inhibition by ebelactone B and phosphoramidon.
International journal of tissue reactions (1995-01-01)
M Saito, M Majima, M Katori, Y Sanjou, I Suyama, H Shiokawa, K Koshiba, T Aoyagi
ABSTRACT
Incubation of bradykinin with human urine resulted in a successive degradation of bradykinin-(1-8), bradykinin-(1-7), bradykinin-(1-6), and bradykinin-(1-5). Although D,L-2-mercaptomethyl-3-guanidinoethylthiopropanoic acid (100 microM) and captopril (100 microM) did not have any significant effect on bradykinin degradation in human urine, the neutral endopeptidase inhibitor phosphoramidon (100 microM), a carboxypeptidase Y-like exopeptidase inhibitor ebelactone B (100 microM), and o-phenanthroline (100 microM) significantly inhibited bradykinin degradation by 36%, 38% and 48% respectively. The combination of phosphoramidon and ebelactone B completely (by 95%) inhibited bradykinin degradation in human urine. At pH 5, bradykinin degradation was performed by carboxypeptidase Y-like exopeptidase; at pH 7, this degradation was performed by neutral endopeptidase in addition to carboxypeptidase Y-like exopeptidase. From these results, it can be concluded that carboxypeptidase Y-like exopeptidase and/or neutral endopeptidase certainly have a role in kinin degradation in human urine under neutral and acid pH conditions.