Merck
  • In vitro primary sensitization of hapten-specific T cells by cultured human epidermal Langerhans cells--a screening predictive assay for contact sensitizers.
In vitro primary sensitization of hapten-specific T cells by cultured human epidermal Langerhans cells--a screening predictive assay for contact sensitizers.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology (1996-05-01)
M Krasteva, J Peguet-Navarro, C Moulon, P Courtellemont, G Redziniak, D Schmitt
ABSTRACT
The need to develop predictive tests which could identify potential allergens has been recognized for many years. There is as yet no accepted in vitro method for the assessment of contact sensitizers. We have tested the ability of a range of contact allergens to induce in vitro primary sensitization of autologous T cells. T-cell proliferation induced by haptens using 2-day cultured human Langerhans cells as antigen-presenting cell was assessed by 3H thymidine incorporation. Antigen specific stimulation was calculated as stimulation indexes. Strong allergens induced in vitro a primary T-cell response in all (trinitrophenyl, TNP: 13/13) or in the majority (fluorescein isothiocyanate, FITC: 7/10) of experiments. An irritant, sodium dodecyl sulfate (SDS), failed to generate a significant T-cell proliferation in any of the experiments (0/10). We obtained a significant lymphoproliferative response to weak sensitizers only in a limited number of experiments: (coumarin: 1/12, citronellal: 0/10, hydroxycitronellal: 2/8). p-Phenylenediamine (PPDA), a prohapten and highly sensitizing chemical in vivo, generated primary sensitization in vitro in only one of six experiments, while Bandrowski's base (BB), a metabolization product of PPDA induced a significant T-cell response in all six experiments. The present in vitro model allows discrimination between two groups of substances: strong contact sensitizers (TNP, FITC, BB) on the one hand and weak sensitizers (coumarin, citronellal and hydroxycitronellal) and irritants (SDS) on the other hand. It could be used as a screening in vitro assay to eliminate strong contact allergens before further predictive animal tests have to be performed.
Sigma-Aldrich
Hydroxycitronellal, ≥95%, FCC, FG