Merck
  • [Synthesis of substituted benzeneseleninic acid and its antineoplastic activity].
[Synthesis of substituted benzeneseleninic acid and its antineoplastic activity].
Hua xi yi ke da xue xue bao = Journal of West China University of Medical Sciences = Huaxi yike daxue xuebao (1995-12-01)
S Zheng, P Xu
ABSTRACT
In view of the negative relationship between selenium and cancer, we designed and synthesized eleven substituted benzeneselenic acids (IVa-k) The antineoplastic activity of the title compounds was evaluated via the tests of inhibition of acetylcholinesterase, HL-60 and K562 in vitro, and Ehrlich carcinoma in mice. The result showed that the compounds IVa and IVb had distinct antineoplastic activity. The life span increase rate of IVb on mice bearing Ehrlich carcinoma was 20.30% (20mg/kg), which was higher than the increase rate of sodium selenite (10.65%, 0.5mg/kg).
Sigma-Aldrich
Benzeneseleninic acid, 99%