Merck
  • Kinetics of p-nitrophenyl pivalate hydrolysis catalysed by cytoplasmic aldehyde dehydrogenase.
Kinetics of p-nitrophenyl pivalate hydrolysis catalysed by cytoplasmic aldehyde dehydrogenase.
The Biochemical journal (1989-01-15)
T M Kitson
ABSTRACT
The effects of modifiers (NAD+, NADH, propionaldehyde, chloral hydrate, diethylstilboestrol and p-nitrobenzaldehyde) on the hydrolysis of p-nitrophenyl (PNP) pivalate (PNP trimethylacetate) catalysed by cytoplasmic aldehyde dehydrogenase are reported. In each case a different inhibition pattern is obtained to that observed when the substrate is PNP acetate; for example, propionaldehyde and chloral hydrate competitively inhibit the hydrolysis of PNP acetate, but are mixed inhibitors with PNP pivalate. The kinetic results can be rationalized in terms of different rate-determining steps: acylation of the enzyme in the case of the pivalate but acyl-enzyme hydrolysis for the acetate. This is confirmed by stopped-flow studies, in which a burst of p-nitrophenoxide is observed when the substrate is PNP acetate, but not when it is the pivalate. PNP pivalate inhibits the dehydrogenase activity of the enzyme competitively with the aldehyde substrate; this is most simply explained if the esterase and dehydrogenase reactions occur at a common enzymic site.
Sigma-Aldrich
4-Nitrophenyl trimethylacetate, 98%