- Mid-infrared optical parametric amplifier based on a LGSe crystal and pumped at 1.6 μm.
Mid-infrared optical parametric amplifier based on a LGSe crystal and pumped at 1.6 μm.
Optics express (2012-12-25)
Etienne Pelletier, Alexander Sell, Alfred Leitenstorfer, R J Dwayne Miller
PMID23262695
ABSTRACT
We demonstrate the generation of 22.6 μJ of combined energy at 3 μm for sub-300fs pulses at a repetition rate of 1 kHz using a LGSe optical parametric amplifier (OPA). The LGSe OPA is pumped by the 140-fs 1.6 μm pulses from a 300-mW KTA optical parametric chirped pulse amplifier (OPCPA) based on an all-optical synchronization scheme. By using a highly-nonlinear fiber, the output of an erbium-doped fiber laser operating at 1560 nm is shifted to 1050 nm in order to coherently seed a Nd:YLF regenerative amplifier. The LGSe OPA is seeded using the MIR coming from the amplification of the 1.6 μm in the OPCPA.