Merck
  • Mazindol in narcolepsy and idiopathic and symptomatic hypersomnia refractory to stimulants: a long-term chart review.
Mazindol in narcolepsy and idiopathic and symptomatic hypersomnia refractory to stimulants: a long-term chart review.
Sleep medicine (2012-10-06)
Nandini Nittur, Eric Konofal, Yves Dauvilliers, Patricia Franco, Smaranda Leu-Semenescu, Valérie Cochen De Cock, Clara O Inocente, Sophie Bayard, Sabine Scholtz, Michel Lecendreux, Isabelle Arnulf
ABSTRACT
Mazindol is a tricyclic, anorectic, non-amphetamine stimulant used in narcolepsy and obesity since 1970. This study aimed to evaluate the long-term benefit/risk ratio in drug-resistant hypersomniacs and cataplexy sufferers. By retrospective analysis of the patients' files in the hospitals of Paris-Salpêtrière (n=91), Montpellier (n=40) and Lyon (n=8), the benefit (Epworth Sleepiness Score (ESS), cataplexy frequency, authorization renewal) and tolerance (side-effects, vital signs, electrocardiogram and cardiac echography) of mazindol were assessed. The 139 patients (45% men) aged 36±15years (range: 9-74) suffered narcolepsy (n=94, 66% with cataplexy), idiopathic (n=37) and symptomatic hypersomnia (n=8) refractory to modafinil, methylphenidate and sodium oxybate. Under mazindol (3.4±1.3mg/day, 1-6mg) for an average of 30months, the ESS decreased from 17.7±3.5 to 12.8±5.1, with an average fall of -4.6±4.7 (p<0.0001) and the frequency of cataplexy fell from 4.6±3.1 to 2±2.8 episodes per week. The cataplexy was eliminated in 14.5% of patients, improved in 27.5%, and unchanged in 29% (missing data in 29%). The treatment was maintained long term in 83 (60%) patients, and stopped because of a lack of efficacy (22%) and/or secondary effects (9%). There was no pulmonary hypertension in the 45 patients who underwent a cardiac echography. The most common adverse effects were dry mouth (13%), palpitations (10%, including one with ventricular hyperexcitability), anorexia (6%), nervousness (6%) and headaches (6%). Mazindol has a long-term, favorable benefit/risk ratio in 60% of drug-resistant hypersomniacs, including a clear benefit on cataplexy.
Sigma-Aldrich
Mazindol, ≥98% (TLC), powder