Merck
  • New insights into the mechanism and an expanded scope of the Fe(III)-mediated vinblastine coupling reaction.
New insights into the mechanism and an expanded scope of the Fe(III)-mediated vinblastine coupling reaction.
Journal of the American Chemical Society (2012-08-04)
Hiroaki Gotoh, Justin E Sears, Albert Eschenmoser, Dale L Boger
ABSTRACT
A definition of the scope of aromatic substrates that participate with catharanthine in an Fe(III)-mediated coupling reaction, an examination of the key structural features of catharanthine required for participation in the reaction, and the development of a generalized indole functionalization reaction that bears little structural relationship to catharanthine itself are detailed. In addition to providing insights into the mechanism of the Fe(III)-mediated coupling reaction of catharanthine with vindoline suggesting the reaction conducted in acidic aqueous buffer may be radical mediated, the studies provide new opportunities for the preparation of previously inaccessible vinblastine analogs and define powerful new methodology for the synthesis of indole-containing natural and unnatural products.
Sigma-Aldrich
Catharanthine, ≥95% (HPLC)