Merck
The pharmacology of McN-A-343.
Pharmacology & therapeutics (2012-05-31)
Frederick J Mitchelson
ABSTRACT
The unusual pharmacology of McN-A-343 was first described by Roszowski in 1961. The agonist appeared to be a selective stimulant of muscarinic receptors in sympathetic ganglia, now known to be the muscarinic M₁ receptor subtype. However, subsequent research demonstrated that McN-A-343 is a partial agonist with similar affinity at all five muscarinic acetylcholine receptor subtypes and its relative selectivity depends on a higher efficacy at the M₁ (and M₄) subtypes. Being a partial agonist its action is also dependent on factors, such as receptor density and coupling efficacy between receptor activation and tissue response. Nevertheless, the relatively high efficacy at M₁ receptors led to its widespread use as an aid to distinguish responses mediated through M₁ receptors from those utilizing M₂ or M₃ muscarinic receptor subtypes, especially in the CNS. There is also evidence that it has an allosteric action at some receptor subtypes. Recently, it was demonstrated that McN-A-343 can bind to an allosteric site on the M₂ receptor as well as to the orthosteric site and has thus been termed a "bitopic agonist". This allosteric site differs from that occupied by allosteric modulators, such as gallamine. Comparison of comparable mutagenic changes in M₂ and M₄ receptors also suggests that McN-A-343 utilizes different regions of the two receptors for ERK1/2 activation. McN-A-343 has a number of non-muscarinic actions. These include activation of some types of nicotinic acetylcholine receptors, antagonism of serotonin 5-HT₃ and 5-HT₄ receptor subtypes, inhibition of the uptake mechanism and a local anesthetic action.
Sigma-Aldrich
McN-A-343, ≥98% (TLC)