Merck
  • Effects of compositional asymmetry in phase behavior of ABA triblock copolymer melts from Monte Carlo simulation.
Effects of compositional asymmetry in phase behavior of ABA triblock copolymer melts from Monte Carlo simulation.
The European physical journal. E, Soft matter (2010-12-02)
S Wołoszczuk, M Banaszak
ABSTRACT
We simulate ABA triblock copolymer melts using a lattice Monte Carlo method, known as cooperative motion algorithm, probing various degrees of compositional asymmetry. Selected order-disorder transition lines are determined in terms of the segment incompatibility, quantified by product χN , and the triblock asymmetry parameters, α and β. We correlate the results of the simulation with the self-consistent field theory and an experimental study of polyisoprene-polystyrene-polyisoprene triblock melt by Hamersky and coworkers. In particular, we confirm the mean-field prediction that for highly asymmetric triblocks the short A -block is localized in the middle of the B -domain due to an entropic advantage. This results in the middle block relaxation and is consistent with the experimental data indicating that as the relatively short A -blocks are grown into AB diblock, from the B -block side, the order-disorder transition temperature is considerably depressed.
Sigma-Aldrich
Negative photoresist I
Pricing and availability is not currently available.