Merck
  • Zolpidem modulation of phasic and tonic GABA currents in the rat dorsal motor nucleus of the vagus.
Zolpidem modulation of phasic and tonic GABA currents in the rat dorsal motor nucleus of the vagus.
Neuropharmacology (2010-03-17)
Hong Gao, Bret N Smith
ABSTRACT
Zolpidem is a widely prescribed sleep aid with relative selectivity for GABA(A) receptors containing alpha1-3 subunits. We examined the effects of zolpidem on the inhibitory currents mediated by GABA(A) receptors using whole-cell patch-clamp recordings from DMV neurons in transverse brainstem slices from rat. Zolpidem prolonged the decay time of mIPSCs and of muscimol-evoked whole-cell GABAergic currents, and it occasionally enhanced the amplitude of mIPSCs. The effects were blocked by flumazenil, a benzodiazepine antagonist. Zolpidem also hyperpolarized the resting membrane potential, with a concomitant decrease in input resistance and action potential firing activity in a subset of cells. Zolpidem did not clearly alter the GABA(A) receptor-mediated tonic current (I(tonic)) under baseline conditions, but after elevating extracellular GABA concentration with nipecotic acid, a non-selective GABA transporter blocker, zolpidem consistently and significantly increased the tonic GABA current. This increase was suppressed by flumazenil and gabazine. These results suggest that alpha1-3 subunits are expressed in synaptic GABA(A) receptors on DMV neurons. The baseline tonic GABA current is likely not mediated by these same low affinity, zolpidem-sensitive GABA(A) receptors. However, when the extracellular GABA concentration is increased, zolpidem-sensitive extrasynaptic GABA(A) receptors containing alpha1-3 subunits contribute to the I(tonic).
Sigma-Aldrich
(R)-(–)-3-Piperidinecarboxylic acid, 97%
Sigma-Aldrich
Nipecotic acid, 98%
Sigma-Aldrich
(S)-(+)-3-Piperidinecarboxylic acid, 97%