Merck
Electrically erodible polymer gel for controlled release of drugs.
Nature (1991-11-28)
I C Kwon, Y H Bae, S W Kim
ABSTRACT
New controlled drug-delivery systems are being explored to overcome the disadvantages of conventional dosage forms. For example, stimulated drug-delivery has been used to overcome the tolerance problems that occur with a constant delivery rate, to mimic the physiological pattern of hormonal concentration and to supply drugs on demand. Stimuli-sensitive polymers, which are potentially useful for pulsed drug delivery, experience changes in either their structure or their chemical properties in response to changes in environmental conditions. Environmental stimuli include temperature, pH, light (ultraviolet or visible), electric field or certain chemicals. Volume changes of stimuli-sensitive gel networks are particularly responsive to external stimuli, but swelling is slow to occur. As well as being useful in the controlled release of drugs, such systems also provide insight into intermolecular interactions. Here we report on a novel polymeric system, which rapidly changes from a solid state to solution in response to small electric currents, by disintegration of the solid polymer complex into two water-soluble polymers. We show that the modulated release of insulin, and by extension other macromolecules, can be achieved with this polymeric system.
Sigma-Aldrich
Poly(2-ethyl-2-oxazoline) 5-7 cSt
Sigma-Aldrich
Poly(2-ethyl-2-oxazoline), average Mw ~500,000, PDI 3‑4
Sigma-Aldrich
Poly(2-ethyl-2-oxazoline), average Mw ~200,000, PDI 3‑4
Sigma-Aldrich
Poly(2-ethyl-2-oxazoline), average Mn 20,000, PDI <1.4