Merck
  • Pharmacokinetics of 5-fluorouracil in elderly Japanese patients with cancer treated with S-1 (a combination of tegafur and dihydropyrimidine dehydrogenase inhibitor 5-chloro-2,4-dihydroxypyridine).
Pharmacokinetics of 5-fluorouracil in elderly Japanese patients with cancer treated with S-1 (a combination of tegafur and dihydropyrimidine dehydrogenase inhibitor 5-chloro-2,4-dihydroxypyridine).
Drug metabolism and disposition: the biological fate of chemicals (2009-04-25)
K Fujita, H Nakayama, W Ichikawa, W Yamamoto, H Endo, F Nagashima, R Tanaka, T Miya, Y Sunakawa, K Yamashita, K Mizuno, H Ishida, K Araki, M Narabayashi, K Miwa, Y Ando, Y Akiyama, K Kawara, T Hirose, Y Sasaki
ABSTRACT
S-1 is an oral anticancer agent that combines tegafur, a prodrug of 5-fluorouracil (5-FU), and 5-chloro-2,4-dihydroxypyridine (CDHP), an inhibitor of dihydropyrimidine dehydrogenase. We examined the effects of aging on the pharmacokinetics of the components of S-1. The median area under the concentration-time curve (AUC) of active 5-FU did not significantly differ between 10 patients 75 years or older and 53 patients younger than 75 years (P = 0.598, Mann-Whitney U test). It is interesting to note that the median oral clearance of tegafur in patients 75 years or older was significantly lower than that in patients younger than 75 years (P = 0.011). Furthermore, the median AUC of CDHP was significantly higher in patients 75 years or older than in those younger than 75 years (P = 0.004). This effect was caused by reduced renal function in the elderly, because CDHP is excreted in the urine by glomerular filtration. The opposing effects of aging on the oral clearance of tegafur and the AUC of CDHP may offset each other, leading to unchanged systemic exposure of 5-FU.
Sigma-Aldrich
2,4-Dihydroxypyridine, 97%