Merck
  • Ground and excited state proton transfer and antioxidant activity of 7-hydroxyflavone in model membranes: absorption and fluorescence spectroscopic studies.
Ground and excited state proton transfer and antioxidant activity of 7-hydroxyflavone in model membranes: absorption and fluorescence spectroscopic studies.
Biophysical chemistry (2008-11-04)
Sudip Chaudhuri, Biswapathik Pahari, Pradeep K Sengupta
ABSTRACT
Steady state and time resolved fluorescence spectroscopy have been used to probe microenvironments of the therapeutically active intrinsically fluorescent flavonoid, 7-hydroxyflavone (7-HF), in model membranes consisting of multilamellar phosphatidylcholine liposomes. Additionally, the antioxidant effects of 7-HF against lipid peroxidation have been evaluated using spectrophotometric assay. Large Stokes shifted emissions with distinct spectroscopic signatures, are observed from the excited state proton transfer (ESPT) tautomer (which is generated by a solvent mediated mechanism) and the ground state anion of 7-HF. The neutral (7-HFN) and anionic (7-HFA) species' appear to be located in the non-polar acyl chain and the polar head group regions of the lipid vesicles respectively. The partition coefficients of 7-HFN and 7-HFA in these vesicles have also been estimated using their intrinsic fluorescence. Anisotropy (r) versus temperature (T) measurements reveal the utility of the tautomer fluorescence anisotropy as a sensitive parameter for exploring structural changes in the membranes. Fluorescence decay kinetics studies indicate heterogeneity in the microenvironments of both 7-HFN and 7-HFA. Furthermore, we demonstrate that lipid peroxidation of the model membranes is partially arrested upon 7-HF binding, suggesting its potential usefulness as an inhibitor of peroxidative damage of cell membranes.
Sigma-Aldrich
7-Hydroxyflavone, ≥98%