Merck
  • Effect of binary combination of some plant-derived molluscicides with MGK-264 or piperonyl butoxide on the reproduction of the snail Lymnaea acuminata.
Effect of binary combination of some plant-derived molluscicides with MGK-264 or piperonyl butoxide on the reproduction of the snail Lymnaea acuminata.
Pest management science (2004-12-28)
Priyamvada Singh, Vinay K Singh, Dinesh K Singh
ABSTRACT
The effects of sub-lethal treatments (20 and 60% of 24-h LC(50)) with plant-derived molluscicides Annona squamosa, acetogenins, Argemone mexicana seed and protopine, in combination (1 + 5) with MGK-264 (ENT 8184) or piperonyl butoxide on the reproduction of Lymnaea acuminata has been studied. The plant-derived molluscicides and their active molluscicidal components, protopine and acetogenins, in combination with ENT 8184 or piperonyl butoxide caused a significant reduction in the fecundity, hatchability and survival of young snails. Combination of A squamosa seed powder with piperonyl butoxide was very effective as it caused a complete arrest of snail fecundity within 24 h of treatment. Removal of the snails to fresh water after the 96-h treatments caused a significant recovery in the fecundity of L acuminata.
Product Description
N-(2-Ethylhexyl)-5-norbornene-2,3-dicarboximide, analytical standard
Supelco
MGK 264, PESTANAL®, analytical standard