Merck
  • The effect of AVP-V receptor stimulation on local GFR in the rat kidney.
The effect of AVP-V receptor stimulation on local GFR in the rat kidney.
Acta physiologica Scandinavica (2004-09-29)
A B Roald, O Tenstad, K Aukland
ABSTRACT
Arginine vasopressin (AVP) might influence urinary concentration ability by altering the intrarenal distribution of glomerular filtration rate (GFR). To study this possibility we have measured the intracortical distribution of GFR following acute AVP-V1 receptor stimulation in anaesthetized female Sprague-Dawley (SPD) rats during euvolemia and water diuresis by the aprotinin method, allowing two consecutive measurements of zonal GFR in the same kidney. Acute i.v. bolus injection of 50 ng V1 receptor agonist ([Phe2, Ile3, Orn8]-vasopressin) followed by a continuous infusion of 5 ng min(-1) in euvolemic rats reduced GFR by 25% in outer cortex (OC), 20% in middle cortex (MC) and 19% in inner cortex (IC) relative to vehicle infusion (all P < 0.05). In water diuretic rats V1 receptor agonist reduced GFR by 22% in OC, 10% in MC and 11% in IC relative to vehicle infusion (P < 0.05). GFR decreased slightly more in OC than in MC and IC in both euvolemic and water diuretic rats (P < 0.05) indicating a distribution of GFR towards MC and IC. Acute infusion of the selective non-peptide V1 receptor antagonist OPC-21268 in euvolemic rats reduced GFR by 14% in OC, 13% in MC and 11% in IC relative to vehicle infusion (P < 0.05), with no significant difference between the layers. The change in distribution of GFR not only between OC and IC, but also between OC and MC suggests that the afferent/efferent arterioles and not the medullary vasa recta is the main site of resistance change. We conclude that acute i.v. infusion of V1 receptor agonist in high doses reduces GFR more in superficial than in deep cortex in both euvolemic and water diuretic rats and that this may be of some importance for water conservation, adding to the V2- receptor effect on water permeability of the collecting ducts.
Sigma-Aldrich
OPC-21268 hydrate, ≥98% (HPLC)