Merck
  • Chemical ecology of oribatid mites III. Chemical composition of oil gland exudates from two oribatid mites, Trhypochthoniellus sp. and Trhypochthonius japonicus (Acari: Trhypochthoniidae).
Chemical ecology of oribatid mites III. Chemical composition of oil gland exudates from two oribatid mites, Trhypochthoniellus sp. and Trhypochthonius japonicus (Acari: Trhypochthoniidae).
Experimental & applied acarology (2003-11-26)
Tomoyo Sakata, Satoshi Shimano, Yasumasa Kuwahara
ABSTRACT
The composition of oil gland exudates from two oribatid mites, Trhypochthoniellus sp. and Trhypochthonius japonicus, was studied with reference to the related species Trhypochthoniellus crassus. Trhypochthoniellus sp. contained a mixture of seven compounds; (Z,Z)-6,9-heptadecadiene, geranial, 3-hydroxybenzene-1,2-dicarbaldehyde (gamma-acaridial), neryl formate, neral, (Z)-8-heptadecene and geranyl formate in decreasing order of abundance. The profile of the components from T. japonicus consisted of two types depending on the locality of sampling with unknown reason; one possessing a mixture of eight compounds [(Z,E)-farnesal, gamma-acaridial, (Z,Z)-6,9-heptadecadiene, (E,E)-farnesal, (Z)-8-heptadecene and geranial in decreasing order] together with two unknown compounds, and the other composed of the same set of compounds together with 2-hydroxy-6-methylbenzaldehyde as the most abundant component. Relative abundance among common components was consistent between the two types of T. japonicus. Profiles of components differed among three species including T. crasus. The phylogenetic relationship between Oribatida and Astigmata was discussed based on secretory compounds commonly distributed between these two suborders.
Sigma-Aldrich
Farnesal, mixture of isomers, technical
Sigma-Aldrich
Farnesal, mixture of isomers, ≥85%