Merck
  • Synthesis of N-acetylglucosaminides of unconjugated and conjugated bile acids.
Synthesis of N-acetylglucosaminides of unconjugated and conjugated bile acids.
Steroids (1992-11-01)
T Niwa, T Koshiyama, J Goto, T Nambara
ABSTRACT
3-N-Acetylglucosaminides of unconjugated, glycine- and taurine-conjugated bile acids have been synthesized. Bile acids appropriately protected were condensed with acetochloroglucosamine through the 3 alpha-hydroxyl group by means of the Koenigs-Knorr reaction using cadmium carbonate as a catalyst. Subsequent borohydride reduction and/or alkaline hydrolysis provided desired 3-N-acetylglucosaminides of unconjugated bile acids. Glycine-conjugates were obtained from N-acetylglucosaminides of unconjugated bile acids and ethyl glycinate by the carbodiimide method. The preparation of N-acetylglucosaminides of taurine-conjugates was attained by the Koenigs-Knorr reaction of bile acid p-nitrophenyl esters followed by condensation with taurine. 7-N-Acetylglucosaminides of ursodeoxycholates were prepared in a similar fashion. The convenient synthesis of 3-N-acetylglucosaminides of unconjugated bile acids is also described.
Sigma-Aldrich
Cadmium carbonate, powder, 98%
Sigma-Aldrich
Cadmium carbonate, powder and chunks, ≥99.996% trace metals basis