Merck
  • Death of cerebellar granule neurons induced by piperine is distinct from that induced by low potassium medium.
Death of cerebellar granule neurons induced by piperine is distinct from that induced by low potassium medium.
Neurochemical research (1998-03-03)
S Unchern, H Saito, N Nishiyama
ABSTRACT
We compared neurotoxicity of piperine and low K+ on cultured cerebellar granule neurons. As considered from lactate dehydrogenase release and 3-(4,5-dimethylthiazol-2-yl)-2,5 diphenyl tetrazolium bromide reduction, both piperine and shifting from high K+ (25 mM) to low K+ (5.4 mM) were cytotoxic to cerebellar granule neurons. Protein synthesis inhibitors, cycloheximide and anisomycin, and an endonuclease inhibitor, aurintricarboxylic acid, were protective against low K+-induced neuronal death whereas they were ineffective against that induced by piperine. D-alpha-tocopherol, trolox, and a spin trap 3,3,5,5-tetramethyl-1-pyrroline-1-oxide were protective against piperine neurotoxicity whereas they had no effect on that induced by low K+. These results suggest that piperine and low K+ may exert neurotoxic effects on cerebellar granule neurons through different mechanisms. Death of cerebellar granule neurons induced by piperine may be mediated by non-apoptotic mechanisms and may involve membrane lipid peroxidation and/or free radical generation.
Sigma-Aldrich
3,3,5,5-Tetramethyl-1-pyrroline N-oxide, 95%