Merck
  • Rapid liquid chromatographic determination of fatty acids as 2-nitrophenylhydrazine derivatives.
Rapid liquid chromatographic determination of fatty acids as 2-nitrophenylhydrazine derivatives.
Journal of AOAC International (1996-03-01)
H Miwa, M Yamamoto
ABSTRACT
Direct derivatization of saponified fats and oils with 2-nitrophenylhydrazine hydrochloride produced corresponding fatty acid hydrazides without time-consuming sample cleanup and/or extraction steps. Hydrazides were injected directly into a liquid chromatograph. Improved isocratic separation was achieved within only 22 min for 29 saturated and mono- and polyunsaturated fatty acids (C8:0-C22:6), including cis-trans isomers and double-bond positional isomers. The technique has good quantitative accuracy while reducing analysis time, cost, and labor.
Sigma-Aldrich
2-Nitrophenylhydrazine, 97%
Pricing and availability is not currently available.