Merck
  • Preparation of 2,3-seco-5 alpha-cholestane-2,3-diol and 4 alpha-methyl-2,3-seco-5 alpha-cholestane-2,3-diol and its reactions with o-nitrophenyl selenocyanate.
Preparation of 2,3-seco-5 alpha-cholestane-2,3-diol and 4 alpha-methyl-2,3-seco-5 alpha-cholestane-2,3-diol and its reactions with o-nitrophenyl selenocyanate.
Steroids (1984-03-01)
M Arnó, M B García, J R Pedro, E Seoane
ABSTRACT
The reaction of 2,3-seco-5 alpha-cholestane-2,3-diol and 4 alpha-methyl-2,3-seco-5 alpha-cholestane-2,3-diol with o-nitrophenyl selenocyanate was studied. The diols were synthesized from cholesterol.
Sigma-Aldrich
2-Nitrophenyl selenocyanate, 97.5%
Pricing and availability is not currently available.