Merck
  • Developmental toxicity of four glycol ethers applied cutaneously to rats.
Developmental toxicity of four glycol ethers applied cutaneously to rats.
Environmental health perspectives (1984-08-01)
B D Hardin, P T Goad, J R Burg
ABSTRACT
Previous NIOSH studies demonstrated the embryo- and fetotoxicity and teratogenicity of ethylene glycol monoethyl ether (EGEE) applied to the shaved skin of pregnant rats. In the present study ethylene glycol monoethyl ether acetate (EGEEA), ethylene glycol monobutyl ether (EGBE), and diethylene glycol monoethyl ether (diEGEE) were tested in the same experimental model, using distilled water as the negative control and EGEE as a positive control. Water or undiluted glycols were applied four times daily on days 7 to 16 of gestation to the shaved interscapular skin with an automatic pipetter. Volumes of EGEE (0.25 mL), EGEEA (0.35 mL), and diEGEE (0.35 mL) were approximately equimolar (2.6 mmole per treatment). EGBE at 0.35 mL four times daily (approximately 2.7 mmole per treatment) killed 10 of 11 treated rats, and was subsequently tested at 0.12 mL (0.9 mmole) per treatment. EGEE- and EGEEA-treated rats showed a reduction in body weight relative to water controls that was associated with completely resorbed litters and significantly fewer live fetuses per litter. Fetal body weights were also significantly reduced in those groups. Visceral malformations and skeletal variations were significantly increased in EGEE and EGEEA groups over the negative control group. No embryotoxic, fetotoxic, or teratogenic effects were detected in the EGBE- or diEGEE-treated litters.
Sigma-Aldrich
2-Ethoxyethyl acetate, 98%
Supelco
2-Ethoxyethyl acetate, certified reference material, TraceCERT®